Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxydiphenyl Sulfone (Technical), MP Biomedicals

Catalog No. ICN205014 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205014 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205014 Supplier MP Biomedicals, Inc Supplier No. 0520501405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00002350
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
PubChem CID 6626
ChEBI CHEBI:34372
IUPAC Name 4-(4-hydroxybenzenesulfonyl)phenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

Specifications

Quantity 100 g
Formula Weight 250.27
Chemical Name or Material 4, 4«-Dihydroxydiphenyl Sulfone, Tech
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.