CAS RN 8002-03-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Promotions Available
Peanut oil
Peanut oil
CAS: 8002-03-7 MDL Number: MFCD00131805 Synonym: Arachis oil,Groundnut oil
2
StatLab MasterTech Peanut Oil
Used as a reagent to protect the acid-fastness of mycobacterial cells during the staining process
3
Fisher Science Education™ Peanut Oil
CAS: 8002-03-7 Synonym: Arachis Oil,Groundnut Oil
4
Peanut Oil, NF, Spectrum™ Chemical
CAS: 8002-03-7