Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Peanut Oil
SCI_ED

Catalog No. S25760 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25760 500 mL Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25760
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 8002-03-7
Synonym Arachis Oil, Groundnut Oil

Specifications

Quantity 500 mL
Assay Percent Range 100% w/v
Packaging Solution
Density 0.9170g/mL
Chemical Name or Material Peanut Oil
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.