accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3NONCH3NHNOCH3OO

CAS RN 8002-89-9

IUPAC Name: sodium 1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione acetate
Synonyms: sodium theophylline acetate
Molecular Weight (g/mol): 262.2
Molecular Formula: C9H11N4NaO4
InChi Key: JDYRVPRLEPMNDB-UHFFFAOYSA-M
SMILES: [Na+].CC([O-])=O.CN1C2=C(NC=N2)C(=O)N(C)C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results