accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNHCl

CAS RN 80028-44-0

IUPAC Name: ethyl pyrrolidine-3-carboxylate hydrochloride
Synonyms: ethyl pyrrolidine-3-carboxylate hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: AYYKCALHTPKNOI-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results