accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNH2NCl

CAS RN 8004-94-2

IUPAC Name: 3-amino-7-(dimethylamino)-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: 3-amino-7-(dimethylamino)-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular Weight (g/mol): 350.85
Molecular Formula: C20H19ClN4
InChi Key: SQHWUYVHKRVCMD-UHFFFAOYSA-N
SMILES: [Cl-].CN(C)C1=CC=C2N=C3C=CC(N)=CC3=[N+](C3=CC=CC=C3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results