Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basic Violet 8, TCI America™
SDP

Catalog No. B47315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47315G Supplier TCI America Supplier No. B47315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent suitable for analytical and research applications.

Chemical Identifiers

CAS 8004-94-2
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00059988
InChI Key SQHWUYVHKRVCMD-UHFFFAOYSA-N
PubChem CID 75442
IUPAC Name 3-amino-7-(dimethylamino)-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(N)=CC3=[N+](C3=CC=CC=C3)C2=C1

Specifications

Quantity 5 g
Physical Form Crystal/Powder
Grade Laboratory
Chemical Name or Material Basic Violet 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.