accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)O(S)OCH3O(R)NNN(R)OCH3O(S)OCH3O

CAS RN 80321-89-7

IUPAC Name: [(2R,3S,4R,5R,6S)-3,4,6-tris(acetyloxy)-5-azidooxan-2-yl]methyl acetate
Synonyms: [(2R,3S,4R,5R,6S)-3,4,6-tris(acetyloxy)-5-azidooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 373.32
Molecular Formula: C14H19N3O9
InChi Key: QKGHBQJLEHAMKJ-DHGKCCLASA-N
SMILES: CC(=O)OC[C@H]1O[C@@H](OC(C)=O)[C@H](N=[N+]=[N-])[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results