accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHOFFFFFF

CAS RN 805239-56-9

IUPAC Name: 4-[2-phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-3-yl]phenol
Synonyms: phtpp
Molecular Weight (g/mol): 423.32
Molecular Formula: C20H11F6N3O
InChi Key: AEZPAUSGTAHLOQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=C2N=C(C=C(N2N=C1C1=CC=CC=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)