accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3CH3CH3H3CH3CH3CCH3CH3H3CCH3

CAS RN 80663-92-9

IUPAC Name: 2,5,8,11-tetra-tert-butylperylene
Synonyms: 2,5,8,11-tetra-tert-butylperylene
Molecular Weight (g/mol): 476.75
Molecular Formula: C36H44
InChi Key: BFTIPCRZWILUIY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=CC(=CC3=C2C(=C1)C1=CC(=CC2=CC(=CC3=C12)C(C)(C)C)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)