Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5,8,11-Tetra-tert-butylperylene 97.0+%, TCI America™
SDP

Catalog No. T3053100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3053100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3053100MG Supplier TCI America Supplier No. T3053100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80663-92-9
Molecular Formula C36H44
Molecular Weight (g/mol) 476.748
MDL Number MFCD09835379
InChI Key BFTIPCRZWILUIY-UHFFFAOYSA-N
Synonym TBP
PubChem CID 15082495
IUPAC Name 2,5,8,11-tetratert-butylperylene
SMILES CC(C)(C)C1=CC2=C3C(=C1)C=C(C=C3C4=CC(=CC5=CC(=CC2=C54)C(C)(C)C)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 312°C
Quantity 100 mg
Formula Weight 476.75
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5,8,11-Tetra-tert-butylperylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.