accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHOH

CAS RN 808-12-8

IUPAC Name: 1,2-bis(4-methylphenyl)-1,2-diphenylethane-1,2-diol
Synonyms: 1,2-bis(4-methylphenyl)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol): 394.51
Molecular Formula: C28H26O2
InChi Key: YOCSNHRJFCUGEU-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results