accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OCH3H3CCH3CH3CH3H3COH3CH3CCH3

CAS RN 809-73-4

IUPAC Name: 2,6-di-tert-butyl-4-[2-(3,5-di-tert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
Synonyms: 2,6-di-tert-butyl-4-[2-(3,5-di-tert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 434.66
Molecular Formula: C30H42O2
InChi Key: VEDXDZRQOYXKTL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results