accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOONH

CAS RN 81-77-6

IUPAC Name: 2,17-diazaheptacyclo[16.12.0.0³,¹⁶.0⁴,¹³.0⁶,¹¹.0¹⁹,²⁸.0²¹,²⁶]triaconta-1(30),3,6,8,10,13,15,18,21,23,25,28-dodecaene-5,12,20,27-tetrone
Synonyms: medium blue C.I. vat blue 4 blue O indanthrone blue
Molecular Weight (g/mol): 442.43
Molecular Formula: C28H14N2O4
InChi Key: UHOKSCJSTAHBSO-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=C3NC4=CC=C5C(=O)C6=CC=CC=C6C(=O)C5=C4NC3=CC=C12
Quantity 
  • (1)
  • (3)
Color 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)