Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indanthrone (Water soluble), TCI America™
SDP

Catalog No. I023025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I023025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I023025G Supplier TCI America Supplier No. I023025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81-77-6
Molecular Formula C28H14N2O4
Molecular Weight (g/mol) 442.43
MDL Number MFCD00046964
InChI Key UHOKSCJSTAHBSO-UHFFFAOYSA-N
PubChem CID 6690
IUPAC Name 2,17-diazaheptacyclo[16.12.0.0³,¹⁶.0⁴,¹³.0⁶,¹¹.0¹⁹,²⁸.0²¹,²⁶]triaconta-1(30),3,6,8,10,13,15,18,21,23,25,28-dodecaene-5,12,20,27-tetrone
SMILES O=C1C2=CC=CC=C2C(=O)C2=C3NC4=CC=C5C(=O)C6=CC=CC=C6C(=O)C5=C4NC3=CC=C12

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 442.43
Physical Form Crystalline Powder
Chemical Name or Material Indanthrone (Water soluble)
EINECSNumber (5)-2227&(5)-3311
RTECSNumber CB8761100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.