Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indanthrone, Water Soluble, Spectrum™ Chemical
SDP

Catalog No. 18620914 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-914 100 g
18-620-913 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-914 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I1276100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Indanthrene, Vat Blue 4, C.I. Pigment Blue 60, Lutetia Fast Blue RS

Chemical Identifiers

CAS 81-77-6

Specifications

Packaging Amber Glass
Color Dark Blue
Quantity 100 g
Chemical Name or Material Indanthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.