accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81029-03-0

IUPAC Name: 1-methoxy-2,3-dimethyl-4-nitrobenzene
Synonyms: 1-methoxy-2,3-dimethyl-4-nitrobenzene
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: DUBFMQLUMHKYOX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=C1C)[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)