Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-p-nitroanisole 98%, Thermo Scientific™

Catalog No. AC198010250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC198010250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC198010250 Supplier Thermo Scientific Chemicals Supplier No. 198010250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81029-03-0
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
InChI Key DUBFMQLUMHKYOX-UHFFFAOYSA-N
Synonym 2,3-dimethyl-4-nitroanisole, 2,3-dimethyl-p-nitroanisole, 4-methoxy-2,3-dimethylnitrobenzene, 3-methoxy-6-nitro-o-xylene, benzene, 1-methoxy-2,3-dimethyl-4-nitro, 1-methoxy-2,3-dimethyl-4-nitro-benzene, pubchem13217, acmc-209pkk, 2,3-dimethy-4-nitrolanisole, ksc496e8f
PubChem CID 596465
IUPAC Name 1-methoxy-2,3-dimethyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1C)OC)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 73°C
Color Beige-Green
Infrared Spectrum Authentic
Linear Formula O2NC6H2(CH3)2OCH3
Quantity 25 g
Formula Weight 181.19
Percent Purity ≥97.5%
Physical Form Chunks and/or Powder
Chemical Name or Material 2, 3-Dimethyl-p-nitroanisole, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.