Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-4-nitroanisole 98.0+%, TCI America™
SDP

Catalog No. D240910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D240910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D240910G Supplier TCI America Supplier No. D240910G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81029-03-0
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD00007163
InChI Key DUBFMQLUMHKYOX-UHFFFAOYSA-N
Synonym 2,3-dimethyl-4-nitroanisole, 2,3-dimethyl-p-nitroanisole, 4-methoxy-2,3-dimethylnitrobenzene, 3-methoxy-6-nitro-o-xylene, benzene, 1-methoxy-2,3-dimethyl-4-nitro, 1-methoxy-2,3-dimethyl-4-nitro-benzene, pubchem13217, acmc-209pkk, 2,3-dimethy-4-nitrolanisole, ksc496e8f
PubChem CID 596465
IUPAC Name 1-methoxy-2,3-dimethyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1C)OC)[N+](=O)[O-]

Specifications

Melting Point 72°C
Color Yellow
Quantity 10 g
Formula Weight 181.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-4-nitroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.