accessibility menu, dialog, popup

UserName

Viewing profile as user:
PbOOOO

CAS RN 811-54-1

IUPAC Name: λ²-lead(2+) diformate
Synonyms: λ²-lead(2+) diformate
Molecular Weight (g/mol): 297.2
Molecular Formula: C2H2O4Pb
InChi Key: ZGVSCHRFRACEIO-UHFFFAOYSA-L
SMILES: [Pb++].[O-]C=O.[O-]C=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results