CAS RN 81167-22-8
IUPAC Name:
2-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-1-ethyl-1H-imidazole hydrochloride
Synonyms:
imiloxan hcl
Molecular Weight (g/mol):
280.75
Molecular Formula:
C14H17ClN2O2
InChi Key:
ANDJPJNFVJXEKX-UHFFFAOYNA-N
SMILES:
Cl.CCN1C=CN=C1CC1COC2=CC=CC=C2O1
1
–
1
of
1
results
1
R&D Systems™ Imiloxan hydrochloride
CAS: 81167-22-8 Molecular Formula: C14H17ClN2O2