accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClBr

CAS RN 81778-09-8

IUPAC Name: 2-(bromomethyl)-1-chloro-4-fluorobenzene
Synonyms: 2-(bromomethyl)-1-chloro-4-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: AUVLFQDKJFSFIX-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(CBr)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)