Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorobenzyl Bromide 96.0+%, TCI America™
SDP

Catalog No. C23595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23595G 5 g
C23591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23595G Supplier TCI America Supplier No. C23595G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81778-09-8
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD01631419
InChI Key AUVLFQDKJFSFIX-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide
PubChem CID 2773623
IUPAC Name 2-(bromomethyl)-1-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)CBr)Cl

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
UN Number 2924
Formula Weight 223.47
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.