Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorobenzyl bromide, 98%, Thermo Scientific™

Catalog No. AAH6125903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6125903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6125903 Supplier Thermo Scientific Chemicals Supplier No. H6125903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81778-09-8
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD01631419
InChI Key AUVLFQDKJFSFIX-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide
PubChem CID 2773623
IUPAC Name 2-(bromomethyl)-1-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)CBr)Cl

Specifications

Color Colorless
Density 1.654 g/mL
Boiling Point 65°C to 66°C
Flash Point 17°C (63°F)
Refractive Index 1.566
Quantity 1 g
UN Number UN2924
Formula Weight 200.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorobenzyl bromide
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.