accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3CH2Br

CAS RN 81790-10-5

IUPAC Name: (2-bromoprop-2-en-1-yl)trimethylsilane
Synonyms: (2-bromoprop-2-en-1-yl)trimethylsilane
Molecular Weight (g/mol): 193.16
Molecular Formula: C6H13BrSi
InChi Key: LUPQCAARZVEFMT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CC(Br)=C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)