CAS RN 81854-56-0
IUPAC Name:
2-(dipropylamino)propanoic acid
Synonyms:
2-(dipropylamino)propanoic acid
Molecular Weight (g/mol):
173.26
Molecular Formula:
C9H19NO2
InChi Key:
GZRZQYMPUHUFSQ-UHFFFAOYNA-N
SMILES:
CCCN(CCC)C(C)C(O)=O
1
–
1
of
1
results
1
N,N-Dipropyl-L-alanine 98.0+%, TCI America™
CAS: 81854-56-0 Molecular Formula: C9H19NO2 Molecular Weight (g/mol): 173.256 MDL Number: MFCD00010823 InChI Key: GZRZQYMPUHUFSQ-QMMMGPOBSA-N PubChem CID: 14274716 IUPAC Name: (2S)-2-(dipropylamino)propanoic acid SMILES: CCCN(CCC)C(C)C(=O)O