accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CBrBrBr

CAS RN 820965-08-0

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium tribromide
Synonyms: 1-butyl-3-methylimidazolium tribromide
Molecular Weight (g/mol): 378.94
Molecular Formula: C8H15Br3N2
InChi Key: RHSQXXVEZYTZHB-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results