accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 821-76-1

IUPAC Name: 1,7-dichloroheptane
Synonyms: 1,7-dichloroheptane
Molecular Weight (g/mol): 169.09
Molecular Formula: C7H14Cl2
InChi Key: PSEMXLIZFGUOGC-UHFFFAOYSA-N
SMILES: ClCCCCCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results