CAS RN 82212-14-4
CAS RN 82212-14-4
IUPAC Name:
4-amino-2-chloro-5-(2H-1,2,3,4-tetrazol-5-yl)benzene-1-sulfonamide
Synonyms:
4-amino-2-chloro-5-(2H-1,2,3,4-tetrazol-5-yl)benzenesulfonamide
Molecular Weight (g/mol):
274.68
Molecular Formula:
C7H7ClN6O2S
InChi Key:
OKHJHVJXPHYALI-UHFFFAOYSA-N
SMILES:
NC1=CC(Cl)=C(C=C1C1=NNN=N1)S(N)(=O)=O
1
–
1
of
1
results
1
5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole 98.0+%, TCI America™
CAS: 82212-14-4 Molecular Formula: C7H7ClN6O2S Molecular Weight (g/mol): 274.683 InChI Key: OKHJHVJXPHYALI-UHFFFAOYSA-N Synonym: 4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulfonamide PubChem CID: 10084756 IUPAC Name: 4-amino-2-chloro-5-(2H-tetrazol-5-yl)benzenesulfonamide SMILES: C1=C(C(=CC(=C1S(=O)(=O)N)Cl)N)C2=NNN=N2