accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82419-36-1

IUPAC Name: 7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Synonyms: ofloxacin tariflox oflodura uro tarivid ofcin ofloxacin, (s)-isomer ofloxacin preparation ofloxacin (product) quinolon oflox
Molecular Weight (g/mol): 361.37
Molecular Formula: C18H20FN3O4
InChi Key: GSDSWSVVBLHKDQ-UHFFFAOYNA-N
SMILES: CC1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCN(C)CC1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (1)
Type 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)