Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ofloxacin, 98%, Thermo Scientific Chemicals

Catalog No. AC455670250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455670250 25 g
AC455670010 1 g
AC455670050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455670250 Supplier Thermo Scientific Chemicals Supplier No. 455670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ofloxacin, CAS # 82419-36-1, is a fluoroquinolone antibiotic with broad antibacterial activity against gram-positive and -negative bacteria, as well as against mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ofloxacin is a synthetic fluoroquinolone antibiotic that is an analog of nalidixic acid
  • Ofloxacin can directly interact with pneumococcal autolysin (LytA) at its choline binding domain, thus inhibiting the ability of LytA to degrade the pneumococcal cell wall
  • Ofloxacin is racemic and its (S)-enantiomer is levofloxacin

Applications

  • Ofloxacin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
  • The DNA and protein-binding capabilities of ofloxacin have been studied in the laboratory
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYSA-N
PubChem CID 4583
ChEBI CHEBI:7731
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Type Ofloxacin
Melting Point 280.0°C to 275.0°C
Infrared Spectrum Authentic
Loss on Drying 0.2% max.
Quantity 25 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 361.37
Percent Purity 98%
Chemical Name or Material Ofloxacin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.