Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ofloxacin, MP Biomedicals™

Catalog No. MP219901805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219901805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219901805 Supplier MP Biomedicals, Inc Supplier No. 0219901805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ofloxacin, White Powder

  • It is used to study pneumococcal cell wall-degrading virulence factors.
  • It is a synthetic fluoroquinolone with broad-spectrum antibacterial activity. It is a nalidixic acid analog.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.37
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYNA-N
Synonym Oflocet, Ofloxacine, Oxaldin, Tarivid

Specifications

Color White
Loss on Drying 0.0002
Quantity 5 g
Solubility Information Soluble in 1 N NaOH(50mg/mL).
Optical Rotation +1 to -1°(Lit.)
Percent Purity 0.9952
Grade Not Available
Physical Form Powder
RTECSNumber UU8815500
storageNote1 Store at Room Temperature(15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.