accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOOONOOOONOOSOHOOO

CAS RN 82436-77-9

IUPAC Name: 1-({8-[(2,5-dioxo-3-sulfopyrrolidin-1-yl)oxy]-8-oxooctanoyl}oxy)-2,5-dioxopyrrolidine-3-sulfonic acid
Synonyms: bissulfosuccinimidyl suberate
Molecular Weight (g/mol): 528.46
Molecular Formula: C16H20N2O14S2
InChi Key: VYLDEYYOISNGST-UHFFFAOYNA-N
SMILES: OS(=O)(=O)C1CC(=O)N(OC(=O)CCCCCCC(=O)ON2C(=O)CC(C2=O)S(O)(=O)=O)C1=O
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results