accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOHSOOO

CAS RN 825-90-1

IUPAC Name: sodium 4-hydroxybenzene-1-sulfonate
Synonyms: sodium phenolsulfonate
Molecular Weight (g/mol): 196.15
Molecular Formula: C6H5NaO4S
InChi Key: BYMHXIQVEAYSJD-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results