Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 4-Hydroxybenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. P0105500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0105500G 500 g
P010525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0105500G Supplier TCI America Supplier No. P0105500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 825-90-1
Molecular Formula C6H5NaO4S
Molecular Weight (g/mol) 196.152
MDL Number MFCD00044734
InChI Key BYMHXIQVEAYSJD-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate
PubChem CID 4379756
IUPAC Name sodium;4-hydroxybenzenesulfonate
SMILES C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 196.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Hydroxybenzenesulfonate
EINECSNumber (3)-1958
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.