accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82560-12-1

IUPAC Name: 5-tert-butyl-1H-pyrazol-3-amine
Synonyms: 5-tert-butyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N)=NN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results