accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82651-72-7

IUPAC Name: 2-[(benzenesulfonyl)methyl]benzonitrile
Synonyms: 2-[(benzenesulfonyl)methyl]benzonitrile
Molecular Weight (g/mol): 257.31
Molecular Formula: C14H11NO2S
InChi Key: MDVRSLRSVHAHNX-UHFFFAOYSA-N
SMILES: O=S(=O)(CC1=CC=CC=C1C#N)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results