Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Phenylsulfonylmethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. P14561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14561G Supplier TCI America Supplier No. P14561G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82651-72-7
Molecular Formula C14H11NO2S
Molecular Weight (g/mol) 257.307
MDL Number MFCD06797158
InChI Key MDVRSLRSVHAHNX-UHFFFAOYSA-N
Synonym 2-(Benzenesulfonylmethyl)benzonitrile, 2-Cyanobenzyl Phenyl Sulfone
PubChem CID 44630459
IUPAC Name 2-(benzenesulfonylmethyl)benzonitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CC2=CC=CC=C2C#N

Specifications

Color White
Quantity 1 g
Formula Weight 257.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Phenylsulfonylmethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.