accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOHCH3CH3H2CNO

CAS RN 82814-77-5

IUPAC Name: 3-hydroxy-2,2,4,4-tetramethyl-5-methylideneimidazolidine-1-carbaldehyde
Synonyms: 3-hydroxy-2,2,4,4-tetramethyl-5-methylideneimidazolidine-1-carbaldehyde
Molecular Weight (g/mol): 184.24
Molecular Formula: C9H16N2O2
InChi Key: VLJZUPMHYLYUJK-UHFFFAOYSA-N
SMILES: CC1(C)N(O)C(C)(C)C(=C)N1C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results