accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82941-26-2

IUPAC Name: 2-butoxyethyl ethaneperoxoate
Synonyms: 2-butoxyethyl ethaneperoxoate
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: OUINRTZUFNPIBX-UHFFFAOYSA-N
SMILES: CCCCOCCOOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results