Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Butoxyethoxy)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. D24915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24915G 5 g
D249125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24915G Supplier TCI America Supplier No. D24915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82941-26-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD01105178
InChI Key OUINRTZUFNPIBX-UHFFFAOYSA-N
Synonym 3,6-Dioxadecanoic Acid
PubChem CID 106312
IUPAC Name 2-butoxyethyl ethaneperoxoate
SMILES CCCCOCCOOC(C)=O

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 g
Formula Weight 176.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (2-Butoxyethoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.