accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-15-8

IUPAC Name: N-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)acetamide
Synonyms: N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide
Molecular Weight (g/mol): 245.28
Molecular Formula: C13H15N3O2
InChi Key: OIAGWXKSCXPNNZ-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C(NC(C)=O)=C1C)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results