Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidoantipyrine 97%, Thermo Scientific™

Catalog No. AC102260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC102260050 Supplier Thermo Scientific Chemicals Supplier No. 102260050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-15-8
Molecular Formula C13H15N3O2
Molecular Weight (g/mol) 245.282
MDL Number MFCD00003141
InChI Key OIAGWXKSCXPNNZ-UHFFFAOYSA-N
Synonym 4-acetamidoantipyrine, 4-acetaminoantipyrine, acetamidoantipyrine, acetylaminoantipyrine, 4-acetoaminoantipyrine, 4-acetylaminophenazone, 4-acetylaminoantipyrine, antipyrine, 4-acetamido, n-acetyl-4-aminoantipyrine, acetylated 4-aminoantipyrine
PubChem CID 65743
ChEBI CHEBI:83513
IUPAC Name N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)NC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Beilstein 24, 274
Formula Weight 245.28
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 4-Acetamidoantipyrine, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.