accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHSOHOO

CAS RN 83-64-7

IUPAC Name: 4-amino-5-hydroxynaphthalene-1-sulfonic acid
Synonyms: 4-amino-5-hydroxynaphthalene-1-sulfonic acid
Molecular Weight (g/mol): 239.25
Molecular Formula: C10H9NO4S
InChi Key: LRDIEHDJWYRVPT-UHFFFAOYSA-N
SMILES: NC1=C2C(O)=CC=CC2=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)