CAS RN 830-13-7
IUPAC Name:
cyclododecanone
Synonyms:
cyclododecanone
Molecular Weight (g/mol):
182.31
Molecular Formula:
C12H22O
InChi Key:
SXVPOSFURRDKBO-UHFFFAOYSA-N
SMILES:
O=C1CCCCCCCCCCC1
1
–
1
of
1
results
1
Cyclododecanone 99.0+%, TCI America™
CAS: 830-13-7 Molecular Formula: C12H22O Molecular Weight (g/mol): 182.307 MDL Number: MFCD00003722 InChI Key: SXVPOSFURRDKBO-UHFFFAOYSA-N Synonym: cyclododecan-1-one,dsstox_cid_7322,dsstox_rid_78407,dsstox_gsid_27322,cyclododecanon,cdon,cyclododecanone,acmc-209pr1,ksc448s4j PubChem CID: 13246 IUPAC Name: cyclododecanone SMILES: C1CCCCCC(=O)CCCCC1