accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNOO

CAS RN 830-74-0

IUPAC Name: 1-(prop-2-en-1-yl)-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 1-(prop-2-en-1-yl)indole-2,3-dione
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: ZWNYDPBLEDGGQD-UHFFFAOYSA-N
SMILES: C=CCN1C(=O)C(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results