accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 831-52-7

IUPAC Name: sodium 2-amino-4,6-dinitrobenzen-1-olate
Synonyms: sodium picramate
Molecular Weight (g/mol): 221.1
Molecular Formula: C6H4N3NaO5
InChi Key: UENNEPPWFZYINW-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC(=CC(=C1[O-])[N+]([O-])=O)[N+]([O-])=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results