Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picramic Acid Sodium Salt, MP Biomedicals™

Catalog No. ICN15625390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15625390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15625390 Supplier MP Biomedicals, Inc Supplier No. 0215625390

missing translation for 'validMainframeMsg'

Picramic Acid Sodium Salt ≥85%, Dark Red Wet Powder

  • Hydrate
  • Purity: 85%
  • Dark red crystals.
  • Contains 20-40% water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-52-7
Molecular Formula C6H4N3O5Na
InChI Key UENNEPPWFZYINW-UHFFFAOYSA-M
Synonym sodium picramate, sodium 2-amino-4,6-dinitrophenolate, sodium 2-amino-4,6-dinitrophenoxide, picramic acid sodium salt, unii-4988szv57r, phenol, 2-amino-4,6-dinitro-, monosodium salt, sdoium picramate wet, 2-amino-4,6-dinitrophenol monosodium salt, picramic acid, sodium salt, 2-amino-4,6-dinitrophenol, sodium salt
PubChem CID 5362461
IUPAC Name sodium;2-amino-4,6-dinitrophenolate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])[O-])N)[N+](=O)[O-].[Na+]

Specifications

Color Red
Flash Point 98.8°C (Lit.)
Assay Percent Range ≥85%
Quantity 500 g
UN Number UN1349
Formula Weight 221.1
Physical Form Solid
Chemical Name or Material Picramic Acid
Grade ≥85%
RTECSNumber TJ7850000
storageNote1 Store at +4°C.
Recommended Storage Store at 2°-8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.