Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picramic Acid Sodium Salt, MP Biomedicals™

Catalog No. MP215625380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215625380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215625380 Supplier MP Biomedicals, Inc Supplier No. 0215625380

missing translation for 'validMainframeMsg'

Picramic Acid Sodium Salt ≥85%, Dark Red Wet Powder

  • Hydrate
  • Purity: 85%
  • Dark red crystals.
  • Contains 20-40% water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-52-7
Molecular Formula C6H4N3NaO5
Molecular Weight (g/mol) 221.10
InChI Key UENNEPPWFZYINW-UHFFFAOYSA-M
Synonym Sodium picramate, Sdoium Picramate Wet

Specifications

Color Dark Red
Flash Point 98.8°C (Lit.)
Quantity 100 g
UN Number UN1349
Physical Form Wet Powder
Grade ≥85%
RTECSNumber TJ7850000
storageNote1 Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.