accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83141-11-1

NH2FOHOF

CAS RN 83141-11-1

IUPAC Name: 3-amino-2,6-difluorobenzoic acid
Synonyms: 3-amino-2,6-difluorobenzoic acid
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: ORSUTLAFOCNIDY-UHFFFAOYSA-N
SMILES: NC1=C(F)C(C(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results